Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109516
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
18
4170
NM_001206856.2
NP_001193785.1
17
4046
NM_001319842.1
NP_001306771.1
17
4084
NM_001319843.1
NP_001306772.1
18
4167
NM_001319844.1
NP_001306773.1
18
3320
NM_001319845.1
NP_001306774.1
19
3425
NM_001319846.1
NP_001306775.1
19
3415
NM_023925.4
NP_076414.2
17
4020
NM_032156.4
NP_115532.3
18
4211
XM_006719138.3
XP_006719201.1
18
4230
XM_006719139.3
XP_006719202.1
17
4188
XM_006719140.3
XP_006719203.1
17
4185
XM_006719142.3
XP_006719205.1
17
4083
XM_006719144.3
XP_006719207.1
16
4041
XM_006719145.3
XP_006719208.1
18
4337
XM_006719146.3
XP_006719209.1
18
4369
XM_006719147.3
XP_006719210.1
18
4274
XM_017019851.1
XP_016875340.1
17
4125
XM_017019852.1
XP_016875341.1
19
3385
XM_017019853.1
XP_016875342.1
19
3455
XM_017019854.1
XP_016875343.1
16
4038
XM_017019855.1
XP_016875344.1
19
3352
XM_017019856.1
XP_016875345.1
18
3310
XM_017019857.1
XP_016875346.1
18
3280
XM_017019858.1
XP_016875347.1
18
3307
XM_017019859.1
XP_016875348.1
17
3136
XM_017019860.1
XP_016875349.1
19
4337
XM_017019861.1
XP_016875350.1
17
3160
XM_017019862.1
XP_016875351.1
19
4334
XM_017019865.1
XP_016875354.1
17
4232
XM_017019866.1
XP_016875355.1
19
3432
XM_017019867.1
XP_016875356.1
19
3459
XM_017019868.1
XP_016875357.1
19
3429
XM_017019869.1
XP_016875358.1
19
3456
XM_017019870.1
XP_016875359.1
19
3369
XM_017019871.1
XP_016875360.1
19
3440
XM_017019872.1
XP_016875361.1
18
3291
XM_017019873.1
XP_016875362.1
17
3207
XM_017019874.1
XP_016875363.1
18
3302
XM_017019875.1
XP_016875364.1
17
3144
XM_017019876.1
XP_016875365.1
16
2853
XM_017019877.1
XP_016875366.1
15
2811
XM_017019878.1
XP_016875367.1
15
2808
XM_017019879.1
XP_016875368.1
17
3137
XM_017019880.1
XP_016875369.1
14
2601
XM_017019881.1
XP_016875370.1
18
3346
XM_017019882.1
XP_016875371.1
18
3286
XM_017019883.1
XP_016875372.1
15
2855
GenBank mRNA
AB058776.1
-
3657
AF525416.1
-
10
1681
AK024247.1
-
5
3136
AK026222.1
-
4
986
AL834212.1
-
16
2698
AY074490.1
-
17
4020
AY074491.1
-
17
4084
BC007520.1
-
4
891
BC066295.1
-
4
922
BC111007.1
-
4
974
BX537569.1
-
15
6226
BX538080.1
-
5
1106
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-9blpc:80/100.66.131.91:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0