Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 7 | 800 | ||
NM_001164392.1 | NP_001157864.1 | 6 | 880 | |||
NM_001164393.1 | NP_001157865.1 | 4 | 599 | |||
NM_007358.3 | NP_031384.1 | 6 | 880 | |||
XM_011541016.1 | XP_011539318.1 | 6 | 836 | |||
XM_011541017.2 | XP_011539319.1 | 6 | 896 | |||
XM_011541019.1 | XP_011539321.1 | 7 | 752 | |||
XM_017000676.1 | XP_016856165.1 | 6 | 735 | |||
XM_017000677.1 | XP_016856166.1 | 6 | 836 | |||
XM_017000678.1 | XP_016856167.1 | 7 | 789 | |||
XM_017000679.1 | XP_016856168.1 | 6 | 836 | |||
GenBank mRNA | AF072814.1 | - | 5 | 578 | ||
AF086908.1 | - | 6 | 805 | |||
AJ010014.1 | - | 6 | 831 | |||
AK290318.1 | - | 6 | 832 | |||
AK302776.1 | - | 7 | 739 | |||
AK302900.1 | - | 4 | 551 | |||
BC001897.1 | - | 6 | 799 | |||
BC010013.2 | - | 6 | 804 | 4308565 |