Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 4 | 931 | ||
NM_006074.4 | NP_006065.2 | 4 | 943 | |||
GenBank mRNA | AK290434.1 | - | 4 | 673 | ||
AK298715.1 | - | 5 | 623 | |||
AK298934.1 | - | 4 | 497 | |||
AK308796.1 | - | 3 | 563 | |||
AK310098.1 | - | 4 | 630 | |||
AL360134.1 | - | 2 | 101 | |||
BC022281.1 | - | 4 | 750 | 4718024 | ||
BC035582.1 | - | 4 | 716 | 5583800 | ||
HQ842635.1 | - | 3 | 564 | |||
X82200.1 | - | 4 | 674 |