Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEACAM4 SIRNA Assays ›
          Gene Symbol:
          CEACAM4
          Gene Name:
          carcinoembryonic antigen related cell adhesion molecule 4
          Gene Aliases:
          CGM7, CGM7_HUMAN, NCA
          Chromosome Location:
          Chr.19: 41616986 - 41627087 on Build GRCh38
          UniGene ID:
          Hs.12
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10524

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEACAM4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001817.3 NP_001808.2 2 266
          XM_006722977.3 XP_006723040.1 2 279
          XM_011526343.1 XP_011524645.1 2 266
          XM_011526345.2 XP_011524647.1 2 279
          XM_011526348.2 XP_011524650.1 2 266
          XM_011526349.2 XP_011524651.1 2 279
          XM_011526350.2 XP_011524652.1 2 266
          XM_011526352.1 XP_011524654.1 2 266
          XM_011526354.1 XP_011524656.1 2 266
          XM_017026199.1 XP_016881688.1 2 266
          XM_017026200.1 XP_016881689.1 2 266
          XM_017026201.1 XP_016881690.1 2 279
          XM_017026202.1 XP_016881691.1 2 279
          XM_017026203.1 XP_016881692.1 2 279
          XM_017026204.1 XP_016881693.1 2 279
          XM_017026205.1 XP_016881694.1 2 266
          XM_017026206.1 XP_016881695.1 2 266
          XM_017026207.1 XP_016881696.1 2 279
          XM_017026208.1 XP_016881697.1 2 279
          XM_017026209.1 XP_016881698.1 2 266
          XM_017026210.1 XP_016881699.1 2 266
          XM_017026211.1 XP_016881700.1 2 266
          XM_017026212.1 XP_016881701.1 2 266
          GenBank mRNA AB924369.1 - 2 155
          AB924370.1 - 2 155
          AK298701.1 - 2 266
          BX644557.1 - 2 267
          BX644579.1 - 2 268
          D90276.1 - 2 252

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          phagocytosis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0