Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYLK SIRNA Assays ›
Gene Symbol:
MYLK
Gene Name:
myosin light chain kinase
Gene Aliases:
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location:
Chr.3: 123612296 - 123884302 on Build GRCh38
UniGene ID:
Hs.477375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103460
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYLK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321309.1
NP_001308238.1
33
5731
NM_053025.3
NP_444253.3
34
5939
NM_053026.3
NP_444254.3
33
5732
NM_053027.3
NP_444255.3
33
5786
NM_053028.3
NP_444256.3
32
5579
XM_011512860.2
XP_011511162.1
31
5657
XM_011512861.2
XP_011511163.1
30
5456
XM_017006469.1
XP_016861958.1
17
3215
XM_017006470.1
XP_016861959.1
16
2203
XM_017006471.1
XP_016861960.1
16
2256
XM_017006472.1
XP_016861961.1
4
549
XM_017006473.1
XP_016861962.1
4
498
GenBank mRNA
AB037663.1
-
17
3015
AF069601.2
-
31
5569
AF069602.1
-
31
5620
AF069603.1
-
30
5413
AF069604.1
-
11
1595
AF096773.1
-
3
573
AF096774.1
-
3
570
AF096775.1
-
3
573
AK127437.1
-
1
1462
AK300610.1
-
16
2189
AK314412.1
-
3
584
AK314443.1
-
3
627
AY339601.1
-
31
5657
AY424269.1
-
30
5452
AY424270.1
-
31
5659
BC017811.1
-
3
594
BC064420.1
-
3
613
BC100761.2
-
3
455
BC100762.1
-
3
455
BC100763.1
-
3
452
BC107783.1
-
3
597
BC113456.1
-
3
418
BC113458.1
-
3
415
CR749364.1
-
3
5399
U48959.2
-
32
5776
X85337.1
-
18
3269
X90870.1
-
9
1289
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
protein phosphorylation
muscle contraction
smooth muscle contraction
tonic smooth muscle contraction
positive regulation of cell migration
bleb assembly
positive regulation of calcium ion transport
aorta smooth muscle tissue morphogenesis
cellular hypotonic response
positive regulation of wound healing
mitophagy in response to mitochondrial depolarization
xenophagy
actin binding
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-775c674fdc-h55f5:80/100.66.134.75:80.
git-commit: 140417879117b66a3780ee196c9dcc5b0ff3ca88
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0