Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300le | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202463.1 | NP_001189392.1 | 2 - 3 | 421 | 117 | |
GenBank mRNA | HQ263411.1 | - | 2 - 3 | 421 | 117 |
Set Membership: |
![]() ![]() ![]() |