Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763466_m1
          See other GPR128 GE Assays ›
          Gene Symbol:
          Gpr128 LOC100911145
          Gene Name:
          G protein-coupled receptor 128
          probable G-protein coupled receptor 128-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.11: 46793777 - 46856291 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 46107178 - 46172303 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01763466_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr128
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107098.1 NP_001100568.1 2351 131
          Gene Symbol: LOC100911145
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751038.3 XP_003751086.1 15 - 16 2179 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000471.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          G protein-coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71912 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157539 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P241690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750768 - -
          Affymetrix Rat Gene 1.0 ST Array 10750779 - -
          Affymetrix Rat Gene 1.0 ST Array 10750767 - -
          Affymetrix Rat Gene 1.0 ST Array 10750770 - -
          Affymetrix Rat Gene 1.0 ST Array 10750780 - -
          Affymetrix Rat Gene 1.0 ST Array 10750777 - -
          Affymetrix Rat Gene 1.0 ST Array 10750766 - -
          Affymetrix Rat Gene 1.0 ST Array 10750778 - -
          Affymetrix Rat Gene 1.0 ST Array 10750776 - -
          Affymetrix Rat Gene 1.0 ST Array 10750774 - -
          Affymetrix Rat Gene 1.0 ST Array 10750771 - -
          Affymetrix Rat Gene 1.0 ST Array 10750769 - -
          Affymetrix Rat Gene 1.0 ST Array 10750773 - -
          Affymetrix Rat Gene 1.0 ST Array 10750765 - -
          Affymetrix Rat Gene 1.0 ST Array 10750775 - -
          Affymetrix Rat Gene 1.0 ST Array 10750772 - -
          Affymetrix Rat Gene 1.1 ST Array 10750755 - -
          Affymetrix Rat Clariom S Assay PSR1100002347.rn.1 TC1100000471.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002346.rn.1 TC1100000471.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10750755 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21929614 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0