Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109269.1 | NP_001102739.1 | 1 - 2 | 108 | 67 | |
XM_008764170.1 | XP_008762392.1 | 1 - 2 | 123 | 67 |
Gene Symbol: LOC503116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240815.2 | XP_006240877.1 | 1 - 2 | 46 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |