Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01640412_s1
          See other DRD5 GE Assays ›
          Gene Symbol:
          Drd5
          Gene Name:
          dopamine receptor D5
          Gene Aliases:
          D1B
          Chromosome Location:
          Chr.14: 77220579 - 77222006 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01640412_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Drd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012768.1 NP_036900.1 1 1359 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Drd5

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic transmission, dopaminergic
          response to amphetamine
          regulation of systemic arterial blood pressure by vasopressin
          norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
          negative regulation of protein kinase activity
          adenylate cyclase-activating G protein-coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          dopamine receptor signaling pathway
          chemical synaptic transmission
          mating behavior
          associative learning
          visual learning
          transmission of nerve impulse
          negative regulation of cell migration
          negative regulation of NAD(P)H oxidase activity
          wound healing
          response to cocaine
          positive regulation of adenylate cyclase activity
          negative regulation of blood pressure
          regulation of female receptivity
          sensitization
          regulation of long-term neuronal synaptic plasticity
          long-term synaptic depression
          cellular response to catecholamine stimulus
          reactive oxygen species metabolic process
          dopamine neurotransmitter receptor activity, coupled via Gs
          G-protein alpha-subunit binding
          dopamine neurotransmitter receptor activity
          protein binding
          dopamine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243283 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006888 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006888 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15525 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777312 - -
          Affymetrix Rat Gene 1.1 ST Array 10777311 - -
          Rat Genome 230 2.0 Array 1369856_at - -
          Rat Genome 230 2.0 Array 1369856_PM_at - -
          Affymetrix HT Rat Focus Array 1369856_at - -
          Affymetrix HT Rat Focus Array 1369856_PM_at - -
          Rat Expression Set 230 1369856_at - -
          Rat Expression Set 230 1369856_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777311 - -
          Rat Genome U34 Set M69118cds_at - -
          Rat Genome U34 Set M69118cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21652345 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368161 - -
          RatRef-12 Expression BeadChip ILMN_54962 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0