Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504973_g1
          See other GRINA GE Assays ›
          Gene Symbol:
          Grina
          Gene Name:
          glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 117304742 - 117307916 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504973_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grina
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153308.4 NP_695220.4 6 - 7 1088 78
          GenBank mRNA BC062074.1 - 6 - 7 1058 78
          S61973.1 - 6 - 7 1038 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001346.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grina

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          endoplasmic reticulum calcium ion homeostasis
          cation transmembrane transport
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          cation channel activity
          transmembrane transporter binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P125659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P125659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P125659 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P125659 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P125659 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702160048 - -
          Agilent Rat cDNA Microarray (G4105A) 702160048 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897150 - -
          Affymetrix Rat Gene 1.0 ST Array 10897151 - -
          Affymetrix Rat Gene 1.0 ST Array 10897153 - -
          Affymetrix Rat Gene 1.0 ST Array 10897149 - -
          Affymetrix Rat Gene 1.0 ST Array 10897147 - -
          Affymetrix Rat Gene 1.0 ST Array 10897152 - -
          Affymetrix Rat Gene 1.0 ST Array 10897148 - -
          Affymetrix Rat Gene 1.1 ST Array 10897146 - -
          Rat Genome 230 2.0 Array 1372724_at - -
          Rat Genome 230 2.0 Array 1372724_PM_at - -
          Affymetrix HT Rat Focus Array 1372724_PM_at - -
          Affymetrix HT Rat Focus Array 1372724_at - -
          Affymetrix Rat Clariom S Assay PSR0700009691.rn.1 TC0700001346.rn.1 RTA TC
          Rat Expression Set 230 1372724_PM_at - -
          Rat Expression Set 230 1372724_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897146 - -
          Rat Genome U34 Set S61973_at - -
          Rat Genome U34 Set S61973_g_at - -
          Rat Genome U34 Set S61973_at - -
          Rat Genome U34 Set S61973_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21195207 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364042 - -
          RatRef-12 Expression BeadChip ILMN_59333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0