Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952731_s1
          See other GM4532 GE Assays ›
          Gene Symbol:
          Gm4532
          Gene Name:
          predicted gene 4532
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 127232418 - 127233130 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952731_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4532
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030674.1 1 451 86
          GenBank mRNA AK011173.1 - 1 453 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY710502.1 - 1 453 Chr. 7 - 127232469 - 127233130 [-] on Build GRCm38 86
          GenBankmRNA BU924688.1 - 1 277 Chr. 7 - 127232423 - 127232955 [-] on Build GRCm38 86
          GenBankmRNA AA041674.1 - 1 210 Chr. 7 - 127232435 - 127232889 [-] on Build GRCm38 86
          GenBankmRNA BX524884.1 - 1 210 Chr. 7 - 127232423 - 127232889 [-] on Build GRCm38 86
          GenBankmRNA BG914766.1 - 1 254 Chr. 7 - 127232430 - 127232898 [-] on Build GRCm38 86
          GenBankmRNA AI427435.1 - 1 Chr. 7 - 127232423 - 127232776 [+] on Build GRCm38 86
          GenBankmRNA AI431057.1 - 1 Chr. 7 - 127232423 - 127232819 [+] on Build GRCm38 86
          GenBankmRNA AI509933.1 - 1 210 Chr. 7 - 127232434 - 127232889 [-] on Build GRCm38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream septin 1 Sept1 Diff6,
          Pnutl3
          Sense Chr. 7 - 127214442 - 127232647 on NCBI Build GRCm38
          Downstream zinc finger protein 553 Zfp553 2600009K23Rik,
          AI431057,
          BC026401,
          C330013F15Rik,
          ENSMUSG00000054461,
          Znf48
          125.0 Anti-sense Chr. 7 - 127233255 - 127238042 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004319.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176025 NR_030674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176025 AK011173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1082736 NR_030674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1082736 NR_030674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180170 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176025 NR_030674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176025 AK011173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1082736 NR_030674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1082736 AK011173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1082736 NR_030674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1082736 NR_030674.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1082736 NR_030674.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180170 - -
          Whole Mouse Genome G4122A A_52_P1082736 NR_030674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1082736 AK011173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1082736 NR_030674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1082736 AK011173.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10568215 - -
          Affymetrix Mouse Clariom S Array PSR0700036285.mm.1 TC0700004319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036285.mm.1 NR_030674.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036285.mm.1 NR_030674.1 Refseq
          Murine Genome U74v2 Set 170380_PM_at - -
          Murine Genome U74v2 Set 170380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821272 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102060603 - -
          Illumina mouse-6 v1.0 expression beadchip 102060603 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218291 - -
          Illumina Sentrix mouse-6.v1 bead array 102060603 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102060603 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218291 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0