Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03049306_m1
          See other ALDH3A1 GE Assays ›
          Gene Symbol:
          Aldh3a1
          Gene Name:
          aldehyde dehydrogenase family 3, subfamily A1
          Gene Aliases:
          Ahd-4, Ahd4, Aldh, Aldh3
          Chromosome Location:
          Chr.11: 61207537 - 61218418 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03049306_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001112725.1 NP_001106196.1 2 - 3 207 96
          NM_007436.2 NP_031462.2 2 - 3 339 96
          GenBank mRNA AF072815.1 - 2 - 3 245 96
          AK144241.1 - 2 - 3 210 96
          BC144946.1 - 2 - 3 239 9053494 96
          BC150677.1 - 2 - 3 239 9087588 96
          EL959277.1 - 2 - 3 195 96
          U12785.1 - 2 - 3 336 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000776.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh3a1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          cellular aldehyde metabolic process
          metabolic process
          positive regulation of cell population proliferation
          response to xenobiotic stimulus
          response to organic cyclic compound
          response to glucocorticoid
          response to cAMP
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD+) activity
          aldehyde dehydrogenase [NAD(P)+] activity
          alcohol dehydrogenase (NADP+) activity
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
          benzaldehyde dehydrogenase (NAD+) activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100006217.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006217.mm.1 NM_001112725.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006217.mm.1 NM_001112725.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006217.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006217.mm.1 NM_007436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006214.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006214.mm.1 NM_001112725.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006214.mm.1 NM_001112725.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006214.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006214.mm.1 NM_007436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006216.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006216.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006216.mm.1 NM_007436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006219.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006219.mm.1 NM_001112725.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006219.mm.1 NM_001112725.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006219.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006219.mm.1 NM_007436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006215.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006215.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006215.mm.1 NM_007436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006218.mm.1 TC1100000776.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006218.mm.1 NM_001112725.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006218.mm.1 NM_001112725.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006218.mm.1 NM_007436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006218.mm.1 NM_007436.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 828804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0