Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01622729_s1
          See other CATSPERG1 GE Assays ›
          Gene Symbol:
          Catsperg1
          Gene Name:
          catsper channel auxiliary subunit gamma 1
          Gene Aliases:
          A230107C01Rik, Catsperg
          Chromosome Location:
          Chr.7: 29181532 - 29215631 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01622729_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsperg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164658.1 NP_001158130.1 28 3361 178
          GenBank mRNA AK133307.1 - 27 3368 178
          BC172039.1 - 21 2342 178

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002717.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK133307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 BC022688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK039202.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK042364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 NM_001164658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK133307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 BC022688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK039202.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 AK042364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P151528 NM_001164658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK133307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 BC022688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK039202.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK042364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 NM_001164658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK133307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 BC022688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK039202.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 AK042364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P151528 NM_001164658.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06285721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10561774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561699 - -
          Affymetrix HT MG-430 PM Array Plate 1425852_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10561690 - -
          Affymetrix Mouse Gene 1.1 ST Array 10561748 - -
          Mouse Genome 430 2.0 Array 1425852_PM_at - -
          Mouse Genome 430 2.0 Array 1425852_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320225_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320225_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700023973.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023973.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023973.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023976.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023976.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023976.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023974.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023974.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023974.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023975.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023975.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023975.mm.1 NM_001164658.1 Refseq
          Mouse Expression Set 430 1425852_PM_at - -
          Mouse Expression Set 430 1425852_at - -
          Mouse Expression Set 430 1445171_PM_at - -
          Mouse Expression Set 430 1445171_at - -
          Murine Genome U74v2 Set 170621_i_at - -
          Murine Genome U74v2 Set 170621_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1425852_PM_at - -
          Mouse Genome 430A 2.0 Array 1425852_at - -
          Affymetrix HT Mouse Genome 430A Array 1425852_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425852_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 870278 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100630064 - -
          Illumina mouse-6 v1.0 expression beadchip 100630064 - -
          Illumina mouse-6 v1.0 expression beadchip 102900377 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212936 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257440 - -
          Illumina Sentrix mouse-6.v1 bead array 102900377 - -
          Illumina Sentrix mouse-6.v1 bead array 100630064 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100630064 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102900377 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2824582 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0