Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145962.2 | NP_666074.1 | 1 - 2 | 454 | 69 | |
GenBank mRNA | AK136305.1 | - | 1 - 2 | 144 | 69 | |
BC027089.1 | - | 1 - 2 | 66 | 5355443 | 69 | |
BC032188.1 | - | 1 - 2 | 500 | 69 | ||
CJ127650.1 | - | 1 - 2 | 357 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |