Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306199_m1
          See other RAB44 GE Assays ›
          Gene Symbol:
          Rab44
          Gene Name:
          RAB44, member RAS oncogene family
          Gene Aliases:
          9830134C10Rik
          Chromosome Location:
          Chr.17: 29114128 - 29148980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306199_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002786.2 NP_001002786.1 3 - 4 2935 86
          GenBank mRNA AK036563.1 - 3 - 4 2936 86
          BC139372.1 - 3 - 4 2543 8862394 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000498.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab44

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          small GTPase mediated signal transduction
          protein transport
          Rab protein signal transduction
          nucleotide binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000409 NM_001002786.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000409 AK036563.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741060 AK081816.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741060 BC010758.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741060 D26123.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741060 NM_007621.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741060 X07411.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44565 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143179 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177554 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000409 NM_001002786.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000409 AK036563.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177554 - -
          Whole Mouse Genome G4122A A_51_P493816 NM_001002786.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P493816 AK036563.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P493816 BC139372.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122338 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143179 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31126180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13426245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13571155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13364898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13426244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13386805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13376484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13071477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13345153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13364901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13403591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13241753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13378753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13559615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13378744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31126156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13345151 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443473 - -
          Affymetrix HT MG-430 PM Array Plate 1456980_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10443470 - -
          Mouse Genome 430 2.0 Array 1456980_at - -
          Mouse Genome 430 2.0 Array 1456980_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 442827_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 442827_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700004382.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004382.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004382.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004384.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004384.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004384.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004388.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004388.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004388.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004385.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004385.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004385.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004387.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004387.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004387.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004386.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004386.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004386.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004389.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004389.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004389.mm.1 NM_001002786.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004383.mm.1 TC1700000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004383.mm.1 NM_001002786.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004383.mm.1 NM_001002786.2 Refseq
          Mouse Expression Set 430 1456980_PM_at - -
          Mouse Expression Set 430 1456980_at - -
          Murine Genome U74v2 Set 129148_at - -
          Murine Genome U74v2 Set 129148_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456980_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456980_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 369798 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102230136 - -
          Illumina mouse-6 v1.0 expression beadchip 102230136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216928 - -
          Illumina Sentrix mouse-6.v1 bead array 102230136 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102230136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3076909 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3155815 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3076909 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3155815 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0