Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264796_g1
          See other GM2176 GE Assays ›
          Gene Symbol:
          Gm2176
          Gene Name:
          glucosamine-6-phosphate deaminase 1 pseudogene
          Gene Aliases:
          A430076E10Rik
          Chromosome Location:
          Chr.18: 80541892 - 80544546 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264796_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2176
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028424.1 3 - 4 1480 115
          GenBank mRNA AK079818.1 - 3 - 4 1482 115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB206501.2 - 1 - 2 400 Chr. 18 - 80541897 - 80542686 [-] on Build GRCm38 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CTD (carboxy-terminal domain,
          RNA polymerase II,
          polypeptide A) phosphatase,
          subunit 1
          Ctdp1 4930563P03Rik,
          AW553592
          -71.3kb Sense Chr. 18 - 80407959 - 80470612 on NCBI Build GRCm38
          Downstream predicted gene,
          41790
          Gm41790 28.8kb Sense Chr. 18 - 80571824 - 80605936 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001662.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101561 NR_028424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101561 AK079818.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63109 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09066419 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101561 NR_028424.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101561 AK079818.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09066419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13781038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14002006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13642247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14085871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13781047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13644290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13644278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14085866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13741840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13644274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31865025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13715523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31864967 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800011901.mm.1 TC1800001662.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011901.mm.1 NR_028424.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 350463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0