Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264668_m1
          See other PSMA8 GE Assays ›
          Gene Symbol:
          Psma8
          Gene Name:
          proteasome (prosome, macropain) subunit, alpha type, 8
          Gene Aliases:
          2410072D24Rik
          Chromosome Location:
          Chr.18: 14706135 - 14762301 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264668_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psma8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163609.1 NP_001157081.1 6 - 7 745 116
          GenBank mRNA AK010717.1 - 6 - 7 746 116
          BC116990.1 - 6 - 7 707 40126309 116
          BC116992.1 - 6 - 7 707 40126311 116
          BC144771.1 - 7 - 8 778 9053319 116
          BC144772.1 - 5 - 6 587 9053320 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000123.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          ubiquitin-dependent protein catabolic process
          proteasomal ubiquitin-independent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          proteolysis involved in cellular protein catabolic process
          endopeptidase activity
          threonine-type endopeptidase activity
          peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514139 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514142 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514145 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P16750 AK010717.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P16750 BC019368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P16750 BC116990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P16750 BC116992.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P16750 BC144771.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867523 AK008226.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867523 AY061970.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867523 BC020084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867523 NM_134029.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492594 AK029934.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61240 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118877 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162568 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P16750 AK010717.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P16750 BC019368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P16750 BC116990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P16750 BC116992.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P16750 BC144771.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03417214 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08919905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08919941 - -
          Whole Mouse Genome G4122A A_51_P213666 AK010717.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213666 BC019368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213666 NM_001163609.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118877 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08919905 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162568 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514142 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514139 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31514144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14055148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13731721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13885943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13976767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13885942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13682237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13621165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31513849 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10454072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454071 - -
          Affymetrix HT MG-430 PM Array Plate 1427170_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10454067 - -
          Mouse Genome 430 2.0 Array 1427170_PM_at - -
          Mouse Genome 430 2.0 Array 1427170_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73677_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73677_at - -
          Affymetrix Mouse Clariom S Array PSR1800000893.mm.1 TC1800000123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000893.mm.1 NM_001163609.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000893.mm.1 NM_001163609.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000892.mm.1 TC1800000123.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000892.mm.1 NM_001163609.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000892.mm.1 NM_001163609.1 Refseq
          Mouse Expression Set 430 1427170_at - -
          Mouse Expression Set 430 1427170_PM_at - -
          Murine Genome U74v2 Set 166354_PM_f_at - -
          Murine Genome U74v2 Set 133945_at - -
          Murine Genome U74v2 Set 166354_f_at - -
          Murine Genome U74v2 Set 133945_PM_at - -
          Mouse Genome 430A 2.0 Array 1427170_at - -
          Mouse Genome 430A 2.0 Array 1427170_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427170_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427170_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757575 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-9blpc:80/100.66.131.91:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0