Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247961_m1
          See other EPPIN GE Assays ›
          Gene Symbol:
          Eppin
          Gene Name:
          epididymal peptidase inhibitor
          Gene Aliases:
          1700024E17Rik, Spinlw1
          Chromosome Location:
          Chr.2: 164588343 - 164593571 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247961_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eppin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029325.2 NP_083601.1 3 - 4 451 60
          GenBank mRNA AF346413.2 - 3 - 4 474 60
          AK006296.1 - 3 - 4 453 60
          BC048637.1 - 3 - 4 474 6771674 60

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eppin

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          molecular_function
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 AF346413.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 AK006296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 NM_029325.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 AF346413.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 AK006296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198053 NM_029325.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543843 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129202 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 AF346413.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 AK006296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 NM_029325.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 AF346413.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 AK006296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198053 NM_029325.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21818754 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129202 - -
          Whole Mouse Genome G4122A A_51_P198053 AF346413.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198053 AK006296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198053 NM_029325.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198053 AF346413.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198053 AK006296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198053 NM_029325.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754857 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01754862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21818708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01978451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02012390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01830588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02108915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02012392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01830597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02150322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01830591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01830599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01754864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818756 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489509 - -
          Affymetrix HT MG-430 PM Array Plate 1420588_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10489506 - -
          Mouse Genome 430 2.0 Array 1420588_PM_at - -
          Mouse Genome 430 2.0 Array 1420588_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75526_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75526_PM_at - -
          Mouse Expression Set 430 1420588_PM_at - -
          Mouse Expression Set 430 1420588_at - -
          Mouse Genome 430A 2.0 Array 1420588_at - -
          Mouse Genome 430A 2.0 Array 1420588_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420588_at - -
          Affymetrix HT Mouse Genome 430A Array 1420588_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 400683 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770706 - -
          Illumina mouse-6 v1.0 expression beadchip 1770706 - -
          Illumina mouse-6 v1.0 expression beadchip 1230487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241630 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661558 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661558 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241630 - -
          mouse-6 v1.1 (Illumina) scl18357.4.1_225-S - -
          mouse-6 v1.1 (Illumina) scl0003097.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230487 - -
          Illumina Sentrix mouse-6.v1 bead array 1770706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003097.1_18-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18357.4.1_225-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770706 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241630 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803415 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661558 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803415 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661558 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241630 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0