Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700022568.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022568.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022568.mm.1 |
NM_024448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022561.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022561.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022561.mm.1 |
NM_024448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022566.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022566.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022566.mm.1 |
NM_024448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022558.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022558.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022558.mm.1 |
NM_024448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022567.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022567.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022567.mm.1 |
NM_024448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022562.mm.1 |
TC1700002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022562.mm.1 |
NM_024448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022562.mm.1 |
NM_024448.2 |
Refseq |