Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238292_s1
          See other GALNT4 GE Assays ›
          Gene Symbol:
          Galnt4
          Gene Name:
          UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4
          Gene Aliases:
          AV011803
          Chromosome Location:
          Chr.10: 99108135 - 99113247 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238292_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galnt4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015737.4 NP_056552.1 1 4981 87
          GenBank mRNA BP761617.1 - 1 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galnt4

          Gene Ontology Categories:

          Process(es)

          polypeptide N-acetylgalactosaminyltransferase activity
          transferase activity
          glycosyltransferase activity
          carbohydrate binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385043 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 BP761617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 AV080565.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 NM_015737.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 BP761617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 AV080565.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229316 NM_015737.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159360 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 BP761617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 AV080565.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 NM_015737.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 BP761617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 AV080565.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229316 NM_015737.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494229 BC057882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494229 U73819.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494229 AK033494.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494229 AK148036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494229 AK153253.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385040 - -
          Whole Mouse Genome G4122A A_52_P229316 BP761617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229316 AV080565.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229316 NM_015737.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229316 BP761617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229316 AV080565.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229316 NM_015737.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494229 BC057882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494229 U73819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494229 AK033494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494229 AK148036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494229 AK153253.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08829088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08279330 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08829089 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08829071 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27385042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27385040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08829091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08829070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08829088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08219624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08279330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08219634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27385052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08829096 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10366041 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366040 - -
          Affymetrix HT MG-430 PM Array Plate 1455915_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423638_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423637_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10366038 - -
          Mouse Genome 430 2.0 Array 1455915_PM_at - -
          Mouse Genome 430 2.0 Array 1423638_at - -
          Mouse Genome 430 2.0 Array 1455915_at - -
          Mouse Genome 430 2.0 Array 1423637_at - -
          Mouse Genome 430 2.0 Array 1423638_PM_at - -
          Mouse Genome 430 2.0 Array 1423637_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14426_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14426_PM_at - -
          Mouse Expression Set 430 1455915_PM_at - -
          Mouse Expression Set 430 1423637_at - -
          Mouse Expression Set 430 1423638_PM_at - -
          Mouse Expression Set 430 1423638_at - -
          Mouse Expression Set 430 1423637_PM_at - -
          Mouse Expression Set 430 1455915_at - -
          Murine Genome U74v2 Set 161016_PM_at - -
          Murine Genome U74v2 Set 161016_at - -
          Murine Genome U74v2 Set 101333_at - -
          Murine Genome U74v2 Set 101333_PM_at - -
          Mouse Genome 430A 2.0 Array 1423637_at - -
          Mouse Genome 430A 2.0 Array 1455915_PM_at - -
          Mouse Genome 430A 2.0 Array 1455915_at - -
          Mouse Genome 430A 2.0 Array 1423638_PM_at - -
          Mouse Genome 430A 2.0 Array 1423637_PM_at - -
          Mouse Genome 430A 2.0 Array 1423638_at - -
          Affymetrix HT Mouse Genome 430A Array 1423638_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455915_at - -
          Affymetrix HT Mouse Genome 430A Array 1455915_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423638_at - -
          Affymetrix HT Mouse Genome 430A Array 1423637_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423637_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2970593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242032 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242032 - -
          mouse-6 v1.1 (Illumina) scl38497.1.1_138-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38497.1.1_138-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242032 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242032 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0