Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190444_g1
          See other ELDR GE Assays ›
          Gene Symbol:
          Eldr
          Gene Name:
          Egfr long non-coding downstream RNA
          Gene Aliases:
          2810442I21Rik, 4930500E12Rik, Fabl
          Chromosome Location:
          Chr.11: 16934709 - 16951282 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190444_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eldr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110421.1 5 - 6 580 73
          NR_110423.1 3 - 4 444 73
          NR_110424.1 3 - 4 427 73
          GenBank mRNA BC054406.1 - 3 - 4 188 73
          BI990615.1 - 5 - 6 380 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002247.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 BC037766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 BC054406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 AK013290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 BC037766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 BC054406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P247927 AK013290.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178106 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 BC037766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 BC054406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 AK013290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 BC037766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 BC054406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P247927 AK013290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P132792 - -
          Whole Mouse Genome G4122A A_52_P247927 BC037766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P247927 BC054406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P247927 AK013290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P247927 BC037766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P247927 BC054406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P247927 AK013290.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27613549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27613561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27613565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09633777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09190564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27613542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27613546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440541_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439112_PM_at - -
          Mouse Genome 430 2.0 Array 1439112_at - -
          Mouse Genome 430 2.0 Array 1439112_PM_at - -
          Mouse Genome 430 2.0 Array 1440541_PM_at - -
          Mouse Genome 430 2.0 Array 1440541_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72735_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72735_at - -
          Affymetrix Mouse Clariom S Array PSR1100021741.mm.1 TC1100002247.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021741.mm.1 NR_110421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021741.mm.1 NR_110423.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021741.mm.1 NR_110424.1 Refseq
          Mouse Expression Set 430 1439112_at - -
          Mouse Expression Set 430 1440541_PM_at - -
          Mouse Expression Set 430 1440541_at - -
          Mouse Expression Set 430 1439112_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440541_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440541_at - -
          Affymetrix HT Mouse Genome 430B Array 1439112_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439112_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 882247 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106110603 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2431292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0