Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01183416_m1
          See other LSM3 GE Assays ›
          Gene Symbol:
          Lsm3
          Gene Name:
          LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae)
          Gene Aliases:
          1010001J12Rik, 2610005D18Rik, 6030401D18Rik, SMX4, USS2
          Chromosome Location:
          Chr.6: 91516035 - 91522620 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01183416_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026309.2 NP_080585.1 3 - 4 258 104
          GenBank mRNA AK003125.1 - 3 - 4 261 104
          AK007849.1 - 3 - 4 250 104
          AK021251.1 - 3 - 4 234 104
          BC054368.1 - 3 - 4 246 5353718 104
          BC062875.1 - 3 - 4 292 5684590 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001088.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm3

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA splicing
          RNA binding
          U6 snRNA 3'-end binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639530 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 BC054368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 BC062875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 AK003125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 AK007849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 BC054368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 BC062875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 AK003125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 AK007849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292056 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155929 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 BC054368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 BC062875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 AK003125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 AK007849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 NM_026309.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 BC054368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 BC062875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 AK003125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 AK007849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 BC054368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 BC062875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 AK003125.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 AK007849.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 BC054368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 BC062875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 AK003125.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 AK007849.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639544 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155929 - -
          Whole Mouse Genome G4122A A_51_P349023 BC054368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 BC062875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 AK003125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 AK007849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 NM_026309.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 BC054368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 BC062875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 AK003125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 AK007849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349023 NM_026309.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138104 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114301 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05115305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05878404 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05878402 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05878403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05426951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05492426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05878404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05492420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05878402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05637102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05294050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05381340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05878403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05778433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05381342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05426950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05708587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10540121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540119 - -
          Affymetrix HT MG-430 PM Array Plate 1448536_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10540118 - -
          Mouse Genome 430 2.0 Array 1448536_at - -
          Mouse Genome 430 2.0 Array 1448536_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67678_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67678_at - -
          Affymetrix Mouse Clariom S Array PSR0600008527.mm.1 TC0600001088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008527.mm.1 NM_026309.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008527.mm.1 NM_026309.2 Refseq
          Mouse Expression Set 430 1448536_PM_at - -
          Mouse Expression Set 430 1448536_at - -
          Murine Genome U74v2 Set 94455_PM_at - -
          Murine Genome U74v2 Set 94455_at - -
          Mouse Genome 430A 2.0 Array 1448536_PM_at - -
          Mouse Genome 430A 2.0 Array 1448536_at - -
          Affymetrix HT Mouse Genome 430A Array 1448536_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448536_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580691 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 780164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252710 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252710 - -
          mouse-6 v1.1 (Illumina) scl067678.4_130-S - -
          Illumina Sentrix mouse-6.v1 bead array 780164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067678.4_130-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2837865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2837865 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067678.4_130-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0