Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008945.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008945.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008945.mm.1 |
NM_001100185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008941.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008941.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008941.mm.1 |
NM_001100185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008942.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008942.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008942.mm.1 |
NM_001100185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008944.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008944.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008944.mm.1 |
NM_001100185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008946.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008946.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008946.mm.1 |
NM_001100185.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008943.mm.1 |
TC0400001224.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008943.mm.1 |
NM_001100185.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008943.mm.1 |
NM_001100185.1 |
Refseq |