Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00816172_s1
          See other CES2G GE Assays ›
          Gene Symbol:
          Ces2g
          Gene Name:
          carboxylesterase 2G
          Gene Aliases:
          2210023G05Rik, Ces2l
          Chromosome Location:
          Chr.8: 104961611 - 104969537 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00816172_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces2g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_197999.2 NP_932116.2 12 2074 90
          GenBank mRNA BC026641.1 - 4 830 90
          BC027185.1 - 12 2052 4194872 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001198.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces2g

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC026641.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC027185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 NM_197999.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 AK146261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC024548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC026641.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC027185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 NM_197999.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 AK146261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P267824 BC024548.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133161 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123001 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114306 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140351 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC026641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC027185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 NM_197999.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 AK146261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC024548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC026641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC027185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 NM_197999.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 AK146261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P267824 BC024548.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26131039 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114306 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123001 - -
          Whole Mouse Genome G4122A A_52_P267824 BC026641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 BC027185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 NM_197999.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 AK146261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 BC024548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 BC026641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 BC027185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 NM_197999.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 AK146261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P267824 BC024548.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121683 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140351 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07431253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07431250 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06994096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06994101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07243152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07079443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06994096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07312344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07077103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06787599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06994094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06909100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07121419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06994100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574581 - -
          Affymetrix HT MG-430 PM Array Plate 1424968_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574572 - -
          Mouse Genome 430 2.0 Array 1424968_at - -
          Mouse Genome 430 2.0 Array 1424968_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72361_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72361_at - -
          Affymetrix Mouse Clariom S Array PSR0800009368.mm.1 TC0800001198.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009368.mm.1 NM_197999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009368.mm.1 NM_197999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009367.mm.1 TC0800001198.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009367.mm.1 NM_197999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009367.mm.1 NM_197999.2 Refseq
          Mouse Expression Set 430 1424968_at - -
          Mouse Expression Set 430 1424968_PM_at - -
          Murine Genome U74v2 Set 104000_at - -
          Murine Genome U74v2 Set 104000_PM_at - -
          Mouse Genome 430A 2.0 Array 1424968_PM_at - -
          Mouse Genome 430A 2.0 Array 1424968_at - -
          Affymetrix HT Mouse Genome 430A Array 1424968_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424968_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 464719 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050136 - -
          Illumina mouse-6 v1.0 expression beadchip 4050136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247906 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247906 - -
          mouse-6 v1.1 (Illumina) scl33429.12_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33429.12_36-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2895284 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247906 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2895284 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247906 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0