Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000452867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
ENST00000452867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000419458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000421505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000452867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
ENST00000452867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016518.hg.1 |
NR_027982.1 |
Refseq |