Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04185566_gH
          See other CTBP2 GE Assays ›
          Gene Symbol:
          CTBP2
          Gene Name:
          C-terminal binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 124987849 - 125161061 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04185566_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTBP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083914.2 NP_001077383.1 3 - 4 493 96
          NM_001290214.2 NP_001277143.1 3 - 4 478 96
          NM_001290215.2 NP_001277144.1 3 - 4 742 96
          NM_001321012.1 NP_001307941.1 3 - 4 437 96
          NM_001321013.1 NP_001307942.1 2 - 3 638 96
          NM_001321014.1 NP_001307943.1 3 - 4 739 96
          NM_001329.3 NP_001320.1 3 - 4 481 96
          XM_005269561.2 XP_005269618.1 4 - 5 664 96
          XM_005269564.2 XP_005269621.1 4 - 5 872 96
          XM_005269567.2 XP_005269624.1 4 - 5 871 96
          XM_005269568.4 XP_005269625.1 4 - 5 599 96
          XM_005269569.2 XP_005269626.1 3 - 4 560 96
          XM_005269571.2 XP_005269628.1 2 - 3 319 96
          XM_005269572.3 XP_005269629.1 2 - 3 367 96
          XM_006717642.2 XP_006717705.1 5 - 6 793 96
          XM_011539351.1 XP_011537653.1 6 - 7 894 96
          XM_011539353.1 XP_011537655.1 5 - 6 765 96
          XM_011539354.1 XP_011537656.1 4 - 5 689 96
          XM_011539355.1 XP_011537657.1 3 - 4 767 96
          XM_017015756.1 XP_016871245.1 4 - 5 535 96
          XM_017015757.1 XP_016871246.1 3 - 4 2261 96
          GenBank mRNA AF016507.1 - 3 - 4 398 96
          AK290390.1 - 3 - 4 342 96
          AL833398.1 - 3 - 4 453 96
          BC002486.2 - 3 - 4 454 96
          BC047018.1 - 3 - 4 458 96
          BC052276.1 - 3 - 4 435 96
          BC072020.1 - 3 - 4 415 96
          BM800106.1 - 1 - 2 153 96
          BQ933925.1 - 474 96
          BT007012.1 - 1 - 2 53 96
          CV029885.1 - 1 - 2 53 96
          CX165167.1 - 3 - 4 307 96
          HY048696.1 - 3 - 4 327 96

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTBP2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          negative regulation of cell population proliferation
          viral genome replication
          positive regulation of chromatin binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          positive regulation of retinoic acid receptor signaling pathway
          white fat cell differentiation
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          protein homodimerization activity
          retinoic acid receptor binding
          NAD binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0