Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03903436_s1
          A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



          Species:
          Human
          Assay Type:
          Non-coding
          Assay ID Hs03903436_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK056250.1 - 1 2154 Chr. 1 - 155231272 - 155233637 [-] on Build GRCh38 71
          GenBankmRNA BQ003930.1 - 1 Chr. 1 - 155231270 - 155231868 [+] on Build GRCh38 71
          GenBankmRNA BU622108.1 - 1 Chr. 1 - 155231270 - 155231802 [+] on Build GRCh38 71
          GenBankmRNA BF992689.1 - 1 Chr. 1 - 155231367 - 155231739 [+] on Build GRCh38 71
          GenBankmRNA R62802.1 - Chr. - - [-] on Build 71
          GenBankmRNA BF993504.1 - 1 Chr. 1 - 155231318 - 155231671 [+] on Build GRCh38 71
          GenBankmRNA AI168161.1 - 1 Chr. 1 - 155231273 - 155231718 [+] on Build GRCh38 71
          GenBankmRNA C05957.1 - Chr. 13 - 44931999 - 44932195 [+] on Build GRCh38 71
          GenBankmRNA N69540.1 - 1 Chr. 1 - 155231270 - 155231732 [+] on Build GRCh38 71
          GenBankmRNA AI080140.1 - 1 Chr. 1 - 155231270 - 155231682 [+] on Build GRCh38 71
          GenBankmRNA DB323781.1 - 1 Chr. 1 - 155231272 - 155231815 [+] on Build GRCh38 71
          GenBankmRNA BI258989.1 - 1 494 Chr. 1 - 155231374 - 155231977 [-] on Build GRCh38 71
          GenBankmRNA BF899341.1 - 1 Chr. 1 - 155231310 - 155231786 [+] on Build GRCh38 71
          GenBankmRNA AA251828.1 - 1 Chr. 1 - 155231272 - 155231663 [+] on Build GRCh38 71
          GenBankmRNA AI264923.1 - 1 Chr. 1 - 155231274 - 155231694 [+] on Build GRCh38 71
          GenBankmRNA BX116933.1 - 1 317 Chr. 1 - 155231270 - 155231800 [-] on Build GRCh38 71
          GenBankmRNA AI693193.1 - 1 Chr. 1 - 155231270 - 155231775 [+] on Build GRCh38 71
          GenBankmRNA AW292308.1 - 1 Chr. 1 - 155231271 - 155231645 [+] on Build GRCh38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glucosylceramidase beta pseudogene 1 GBAP1 GBAP -3.7kb Sense Chr. 1 - 155213825 - 155227534 on NCBI Build GRCh38
          Downstream glucosylceramidase beta GBA GBA1,
          GCB,
          GLUC
          811.0 Sense Chr. 1 - 155234448 - 155244862 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
          git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0