Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671605_s1
          See other GSN-AS1 GE Assays ›
          Gene Symbol:
          GSN-AS1
          Gene Name:
          GSN antisense RNA 1
          Gene Aliases:
          C9orf31, MOST2
          Chromosome Location:
          Chr.9: 121280768 - 121285530 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671605_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GSN-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_103560.1 1 4669 71
          GenBank mRNA AF220263.1 - 1 4649 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RAB14,
          member RAS oncogene family
          RAB14 FBP,
          RAB-14
          -78.7kb Sense Chr. 9 - 121178137 - 121202087 on NCBI Build GRCh38
          Downstream uncharacterized LOC105376256 LOC105376256 6.7kb Sense Chr. 9 - 121292216 - 121299157 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011399.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P367663 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P367663 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P94656 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 42848 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 2802 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26491 - -
          SurePrint G3 Human GE Microarray G4851A ERCC-00109_120 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P94656 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P367663 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P94656 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040688_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040687_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040686_at - -
          Affymetrix Human Gene 1.0 ST Array 8163894 - -
          Affymetrix Human Gene 1.0 ST Array 8163895 - -
          Affymetrix Human Gene 1.0 ST Array 8163893 - -
          Affymetrix Human Gene 1.1 ST Array 8163892 - -
          Affymetrix HT HG-U133+ PM Array Plate 221072_PM_at - -
          Affymetrix Human Genome U219 Array 11762372_at - -
          Affymetrix Human Genome U219 Array 11762372_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 TC0900011399.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177774.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177772.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177772.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177772.hg.1 TC0900011399.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177772.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177772.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177775.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177775.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177775.hg.1 TC0900011399.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177775.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177775.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177773.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177773.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900177773.hg.1 TC0900011399.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177773.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177773.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000414544.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 ENST00000437135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 NR_103560.1 Refseq
          Affymetrix Human Clariom D Assay TC0900011399.hg.1 NR_103560.1 Refseq
          Human Genome U133 Plus 2.0 Array 221072_at - -
          Human Genome U133 Plus 2.0 Array 221072_PM_at - -
          Human Genome U133A 2.0 Array 221072_PM_at - -
          Human Genome U133A 2.0 Array 221072_at - -
          AffymetrixPathwork Diagnostics Pathchip pw574799 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221072_at9 - -
          Human Genome U133 Set 221072_PM_at - -
          Human Genome U133 Set 221072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 116837 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1750173 AF220263.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750173 AF220263.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750173 AF220263.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750173 AF220263.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750173 AF220263.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0