Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03004792_mH
          See other TBX20 GE Assays ›
          Gene Symbol:
          TBX20
          Gene Name:
          T-box 20
          Gene Aliases:
          ASD4
          Chromosome Location:
          Chr.7: 35199936 - 35254100 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03004792_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBX20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077653.2 NP_001071121.1 2 - 3 857 60
          NM_001166220.1 NP_001159692.1 2 - 3 857 60
          GenBank mRNA AJ237589.1 - 1 - 2 89 60
          BC120945.1 - 2 - 3 388 60
          BC120946.1 - 2 - 3 388 40115117 60
          DQ986374.1 - 2 - 3 904 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010719.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001276.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          branching involved in blood vessel morphogenesis
          endoderm formation
          neuron migration
          heart looping
          embryonic heart tube morphogenesis
          outflow tract septum morphogenesis
          tricuspid valve development
          aortic valve morphogenesis
          pulmonary valve formation
          endocardial cushion morphogenesis
          cardiac chamber formation
          cardiac right ventricle morphogenesis
          endocardial cushion formation
          cardiac septum development
          pericardium morphogenesis
          transcription by RNA polymerase II
          muscle contraction
          blood circulation
          cell population proliferation
          dorsal/ventral pattern formation
          negative regulation of SMAD protein complex assembly
          visceral motor neuron differentiation
          foramen ovale closure
          embryonic heart tube elongation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          lateral mesoderm formation
          cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          atrial septum morphogenesis
          pulmonary vein morphogenesis
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          transcription coactivator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07019863.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07019863.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07019863.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07019863.hg.1 TC07001276.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010719.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 NM_001166220.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 TC0700010719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 ENST00000408931.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 ENST00000492961.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 NM_001077653.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700176374.hg.1 NM_001166220.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0