Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02576335_s1
          See other REP15 GE Assays ›
          Gene Symbol:
          REP15
          Gene Name:
          RAB15 effector protein
          Gene Aliases:
          RAB15EP
          Chromosome Location:
          Chr.12: 27696495 - 27697633 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02576335_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: REP15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029874.1 NP_001025045.1 1 847 155

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007157.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000268.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene REP15

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor recycling
          positive regulation of defense response to virus by host
          transferrin transport
          positive regulation of mitophagy in response to mitochondrial depolarization
          xenophagy
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16543248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16543245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08266517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28698474 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3247624 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1144 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1145 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9088 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02593241 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3247624 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3247624 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266501 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16543245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266507 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266498 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698456 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266507 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266501 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266498 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16543245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08266505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28698456 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266504 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266498 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266503 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266509 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266506 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266502 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266501 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266495 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266494 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08266492 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P39624329 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39624329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266526 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266509 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266492 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266506 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266525 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266528 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543250 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16543245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266505 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08266494 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39624329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001029874_at - -
          Affymetrix Human Gene 1.0 ST Array 7954590 - -
          Affymetrix Human Gene 1.1 ST Array 7954589 - -
          Affymetrix Human Genome U219 Array 11740632_at - -
          Affymetrix Human Genome U219 Array 11740632_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12003289.hg.1 ENST00000310791.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12003289.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12003289.hg.1 TC12000268.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12003290.hg.1 ENST00000310791.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12003290.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12003290.hg.1 TC12000268.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200150281.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150281.hg.1 TC1200007157.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200150281.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150279.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150279.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150279.hg.1 TC1200007157.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200150279.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150279.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150278.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150278.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150278.hg.1 TC1200007157.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200150278.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150278.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150277.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150277.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150277.hg.1 TC1200007157.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200150277.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150277.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150280.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150280.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200150280.hg.1 TC1200007157.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200150280.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200150280.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 ENST00000310791.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 NM_001029874.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007157.hg.1 NM_001029874.2 Refseq
          Human Genome U95 Set 66874_PM_r_at - -
          Human Genome U95 Set 66874_r_at - -
          Human Genome U95 Set 90276_at - -
          Human Genome U95 Set 90276_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048469 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A048469 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 166743 - -
          ABI Human Genome Survey Microarray Version 2.0 182760 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1665884 NM_001029874.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1665884 AY662682.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673817 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_26617-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1673817 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1673817 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 3390010 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_26617 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1665884 BC140921.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1673817 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665884 NM_001029874.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665884 AY662682.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1665884 BC140921.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0