Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01104194_m1
          See other CSF2RA GE Assays ›
          Gene Symbol:
          CSF2RA
          Gene Name:
          colony stimulating factor 2 receptor alpha subunit
          Gene Aliases:
          CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
          Chromosome Location:
          Chr.X: 1268800 - 1325097 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01104194_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CSF2RA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161529.1 NP_001155001.1 11 - 12 1268 66
          NM_001161531.1 NP_001155003.1 10 - 11 1140 66
          NM_001161532.1 NP_001155004.1 8 - 9 909 66
          NM_006140.4 NP_006131.2 10 - 11 1140 66
          NM_172245.2 NP_758448.1 10 - 11 1116 66
          XM_011546165.2 XP_011544467.1 9 - 10 1018 66
          XM_011546166.1 XP_011544468.1 9 - 10 1018 66
          XM_011546167.2 XP_011544469.1 9 - 10 1018 66
          XM_011546168.2 XP_011544470.1 9 - 10 1018 66
          XM_011546169.1 XP_011544471.1 9 - 10 1018 66
          XM_011546170.2 XP_011544472.1 9 - 10 1018 66
          XM_017029287.1 XP_016884776.1 10 - 11 1130 66
          GenBank mRNA AK293086.1 - 10 - 11 1098 66
          AK301395.1 - 8 - 9 891 66
          AL833597.1 - 10 - 11 1077 66
          BC002635.1 - 10 - 11 1116 3606186 66
          BC071835.1 - 11 - 12 1238 4650755 66
          M64445.1 - 10 - 11 1108 66
          X17648.1 - 10 - 11 1095 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00006446.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CSF2RA

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          peptidyl-tyrosine phosphorylation
          granulocyte colony-stimulating factor signaling pathway
          positive regulation of GTPase activity
          cellular protein metabolic process
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          protein tyrosine kinase activity
          granulocyte colony-stimulating factor receptor activity
          protein binding
          signaling receptor activity
          granulocyte colony-stimulating factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381509.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381524.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381529.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000417535.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000432318.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000486791.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000498153.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000501036.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161529.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161530.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161531.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_006140.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_172245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000432318.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 TC0X00006446.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381509.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381524.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000381529.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000417535.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000486791.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000498153.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 ENST00000501036.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161529.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161530.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161531.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_001161532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_172245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00144180.hg.1 NM_006140.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0