Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01041121_g1
          See other RHOA GE Assays ›
          Gene Symbol:
          RHOA
          Gene Name:
          ras homolog family member A
          Gene Aliases:
          ARH12, ARHA, RHO12, RHOH12
          Chromosome Location:
          Chr.3: 49359136 - 49412097 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01041121_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RHOA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001313941.1 NP_001300870.1 4 - 5 649 114
          NM_001313943.1 NP_001300872.1 3 - 4 554 114
          NM_001313944.1 NP_001300873.1 3 - 4 494 114
          NM_001313945.1 NP_001300874.1 3 - 4 431 114
          NM_001664.3 NP_001655.1 3 - 4 554 114
          GenBank mRNA AF498970.1 - 2 - 3 274 114
          AK130066.1 - 3 - 4 550 114
          AK130808.1 - 3 - 4 430 114
          AK222556.1 - 3 - 4 446 114
          AK296654.1 - 3 - 4 370 114
          AK307220.1 - 3 - 4 329 114
          BC001360.2 - 3 - 4 386 3051460 114
          BC005976.1 - 3 - 4 391 4102976 114
          BQ721697.1 - 3 - 4 427 114
          BT019870.1 - 2 - 3 274 114
          BX647063.1 - 3 - 4 450 114
          L09159.1 - 2 - 3 263 114
          L25080.1 - 3 - 4 425 114
          X05026.1 - 3 - 4 432 114

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001313945.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RHOA

          Gene Ontology Categories:

          Function(s) Process(es)

          transforming growth factor beta receptor signaling pathway
          Rho protein signal transduction
          viral process
          cell migration
          substantia nigra development
          actin cytoskeleton organization
          platelet activation
          regulation of cell migration
          actin cytoskeleton reorganization
          positive regulation of cytokinesis
          regulation of actin cytoskeleton organization
          regulation of osteoblast proliferation
          Roundabout signaling pathway
          cleavage furrow formation
          apolipoprotein A-I-mediated signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          stress fiber assembly
          apical junction assembly
          endothelial cell migration
          ossification involved in bone maturation
          wound healing, spreading of cells
          positive regulation of neuron differentiation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          negative regulation of axonogenesis
          positive regulation of axonogenesis
          negative chemotaxis
          regulation of small GTPase mediated signal transduction
          positive regulation of stress fiber assembly
          Wnt signaling pathway, planar cell polarity pathway
          positive regulation of lipase activity
          trabecula morphogenesis
          positive regulation of protein serine/threonine kinase activity
          negative regulation of cell migration involved in sprouting angiogenesis
          mitotic spindle assembly
          endothelial tube lumen extension
          positive regulation of NIK/NF-kappaB signaling
          skeletal muscle satellite cell migration
          mitotic cleavage furrow formation
          regulation of cell motility
          GTPase activity
          protein binding
          GTP binding
          myosin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 TC0300011060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300181920.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000265538.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000418115.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000422781.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000431929.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000445425.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 ENST00000454011.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq
          Affymetrix Human Clariom D Assay TC0300011060.hg.1 NM_001664.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0