Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001891.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001891.hg.1 |
ENST00000513449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001891.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001891.hg.1 |
TC04000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000504134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000504134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
TC0400006756.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146869.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000389737.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000504134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000504134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000511366.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000513449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000528167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000531038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
ENST00000532937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006756.hg.1 |
NM_152544.2 |
Refseq |