Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00380520_m1
          See other ATP6V0E2-AS1 GE Assays ›
          Gene Symbol:
          ATP6V0E2-AS1
          Gene Name:
          ATP6V0E2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 149867694 - 149873862 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00380520_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP6V0E2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC028735.2 - 2 - 3 770 91

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 858493 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 312617 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 50654 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 412959 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P146659 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4953 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4952 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4456 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 39219 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P146659 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P146659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866934 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130088 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130088 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866934 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26130097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05866958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16034982 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P146659 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P146659 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32172972 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32173022 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01834420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866933 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866940 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866880 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16034983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866945 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866932 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866947 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16034975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866949 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866934 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866931 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866935 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16034982 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866956 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16034986 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05866942 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01834420 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8143764 - -
          Affymetrix Human Gene 1.0 ST Array 8143763 - -
          Affymetrix Human Gene 1.0 ST Array 8143760 - -
          Affymetrix Human Gene 1.0 ST Array 8143761 - -
          Affymetrix Human Gene 1.0 ST Array 8143765 - -
          Affymetrix Human Gene 1.0 ST Array 8143762 - -
          Affymetrix Human Gene 1.1 ST Array 8143759 - -
          Affymetrix HT HG-U133+ PM Array Plate 229094_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000461019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000464939.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 ENST00000488315.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013634.hg.1 NR_027040.1 Refseq
          Human Genome U133 Plus 2.0 Array 229094_PM_at - -
          Human Genome U133 Plus 2.0 Array 229094_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229094_at3 - -
          Human Genome U95 Set 46589_at - -
          Human Genome U95 Set 46589_PM_at - -
          Human Genome U133 Set 229094_PM_at - -
          Human Genome U133 Set 229094_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033727 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033727 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227240 - -
          ABI Human Genome Survey Microarray Version 2.0 235123 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2067682 BC023225.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2067682 BC028735.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2067682 NR_027040.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2067682 AW452405.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1730662 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2067682 BC023225.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2067682 BC028735.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2067682 NR_027040.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2067682 AW452405.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1730662 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1730662 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2067682 BC023225.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2067682 BC028735.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2067682 NR_027040.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2067682 AW452405.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2067682 BC023225.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2067682 BC028735.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2067682 NR_027040.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2067682 AW452405.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2067682 BC023225.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2067682 BC028735.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2067682 NR_027040.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2067682 AW452405.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2067682 BC023225.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2067682 BC028735.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2067682 NR_027040.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2067682 AW452405.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0