Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00270424_m1
          See other CCNB2 GE Assays ›
          Gene Symbol:
          CCNB2
          Gene Name:
          cyclin B2
          Gene Aliases:
          HsT17299
          Chromosome Location:
          Chr.15: 59105085 - 59125045 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00270424_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCNB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004701.3 NP_004692.1 1 - 2 210 73
          GenBank mRNA AB020981.1 - 1 - 2 95 73
          AF002822.1 - 1 - 2 45 73
          AF087910.1 - 1 - 2 152 73
          AK001404.1 - 1 - 2 174 73
          AK222611.1 - 1 - 2 143 73
          AL080146.1 - 1 - 2 163 73
          BC105086.1 - 1 - 2 158 8144089 73
          BC105112.1 - 1 - 2 158 8144115 73
          BU934242.1 - 1 - 2 211 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007392.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000449.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB020981.1,AF002822.1,AF087910.1,AK001404.1,AL080146.1,BC105086.1,BC105112.1,NM_004701.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human DNA Damage Induced 14-3-3 Sigma Signaling HUMAN CYCLINS AND CELL CYCLE REGULATION 1700 Inventoried MicroArray Quality Control (MAQC) Human_DNA_Damage_Induced_14-3-3_Sigma_Signaling Human_Cyclins_and_Cell_Cycle_Regulation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCNB2

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          in utero embryonic development
          mitotic nuclear division
          mitotic nuclear envelope disassembly
          growth
          T cell homeostasis
          thymus development
          cell division
          regulation of cell cycle
          cell-cell adhesion
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15004274.hg.1 ENST00000288207.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004274.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004274.hg.1 TC15000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15004277.hg.1 ENST00000288207.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004277.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004277.hg.1 TC15000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700010251.hg.1 ENST00000399411.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004276.hg.1 ENST00000288207.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004276.hg.1 ENST00000559622.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004276.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004276.hg.1 TC15000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152642.hg.1 TC1500007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700010251.hg.1 ENST00000399411.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152637.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152637.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152637.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152637.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152637.hg.1 TC1500007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500152640.hg.1 TC1500007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000288207.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559301.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000559622.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000561077.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 ENST00000621385.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Affymetrix Human Clariom D Assay TC1500007392.hg.1 NM_004701.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 169571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0