Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200363.1 | NP_956657.1 | 4 - 5 | 1434 | 64 | |
GenBank mRNA | BC053201.1 | - | 4 - 5 | 1434 | 64 | |
BC165840.1 | - | 3 - 4 | 1167 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |