Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314137.1 | NP_001301066.1 | 943 | 73 | ||
XM_014116307.2 | XP_013971782.1 | 1156 | 73 | |||
GenBank mRNA | AB679832.1 | - | 943 | 73 |
Set Membership: |
![]() ![]() ![]() |