Smart-Vue™ Ancillary Parts
Product Image
Thermo Scientific™

Smart-Vue™ Ancillary Parts

Choose from an assortment of ancillary parts including power supplies, cables and mounting hardware.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludes
SV502-500-LSBTemperature Flat Cable
SV501-500-LSBPower supply, 100/240V to 12 VDC for audio/visual alarm device
SV501-501-LSBPower supply, 100/240V to 6 VDC for CO2 systemFour inter-changeable wall plug configurations
SV501-502-LSBPower supply, 100/240V to 5 VDC for repeater or receiverFour inter-changeable wall plug configurations
SV502-501-LSBTemperature/RH Flat Cable
SV503-500-LSB3.6V Lithium Battery
SV504-500-LSB
SV505-500-LSBSensor Mounting Kits
SV506-500-LSBBlue Rubber Knob; Pack of 10
SV507-500-LSB
SV508-500-LSBWall Mounting Holder for TCP/IP Receiver or Repeaters
SV510-500-LSBReplacement Battery Cover
7203FMS4-20 milliamp output for use with Smart-Vue wireless monitoring solution on FMS high-performance flammable storage freezers
Catalog number SV502-500-LSB
Price (USD)
-
Request A Quote
Description:
Temperature Flat Cable

Choose from an assortment of Thermo Scientific™ Smart-Vue™ Ancillary Parts, including power supplies, cables and mounting hardware for use with Smart-Vue Wireless Monitoring Systems.

Compatible With:

  • Refrigerators
  • Freezers
  • LN2 Storage
  • CO2 Incubators
  • Clean Rooms
  • Storage Areas
  • Ovens

Ordering Information:

Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.

Specifications
TypeCable
DescriptionTemperature Flat Cable
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application)Cold Storage
For Use With (Equipment)Smart-Vue™ Wireless Monitoring Systems
Unit SizeEach