Ebselen, Thermo Scientific Chemicals
Ebselen, Thermo Scientific Chemicals
Ebselen, Thermo Scientific Chemicals
Ebselen, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ebselen, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J63190.MA
also known as J63190-MA
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63190.MA
also known as J63190-MA
Price (USD)
106.00
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)
106.00
Each
Add to cart
Chemical Identifiers
CAS60940-34-3
IUPAC Name2-phenyl-2,3-dihydro-1,2-benzoselenazol-3-one
Molecular FormulaC13H9NOSe
InChI KeyDYEFUKCXAQOFHX-UHFFFAOYSA-N
SMILESO=C1N([Se]C2=CC=CC=C12)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow to green
Melting Point (clear melt)179-184°C
Assay from Supplier's CofA≥97.5%
Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

Solubility
Soluble in ethanol, methanol, acetone, acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Helmut Sies. Ebselen, a selenoorganic compound as glutathione peroxidase mimic. Free Radical Biology and Medicine. 1993, 14,(3), 313-323.
  2. Tankred Schewe. Molecular actions of Ebselen—an anti-inflammatory antioxidant. General Pharmacology: The Vascular System. 1995, 26,(6), 1153-1169.