Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO
Note: Product 87729 will be discontinued and replaced by product 66383, 66384, or 88404 by 6/30/23.
Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO
Thermo Scientific™

Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation, self-floating protein dialysis cassettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSample Volume (Metric)
87729G2 Dialysis Cassettes0.5 mL
87730G2 Dialysis Cassettes3 mL
87732G2 Dialysis Cassettes30 mL
88250G2 Dialysis Cassettes (gamma-irradiated)0.5 mL
88251G2 Dialysis Cassettes (gamma-irradiated)3 mL
88252G2 Dialysis Cassettes (gamma-irradiated)15 mL
88253G2 Dialysis Cassettes (gamma-irradiated)30 mL
88254G2 Dialysis Cassettes (gamma-irradiated)70 mL
Catalog number 87729
Price (USD)
147.00
Each
Add to cart
Description:
G2 Dialysis Cassettes
Sample Volume (Metric):
0.5 mL
Request bulk or custom format
Price (USD)
147.00
Each
Add to cart
Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis. They allow samples to be loaded with a syringe or pipette through the unique sample port. The sample is easily recovered through either the pipette or syringe sample ports. The Slide-A-Lyzer Cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems.
Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

The 10K molecular-weight cutoff (MWCO) cassettes use a low-binding regenerated cellulose that is rated to completely retain proteins and other macromolecules larger than 10,000 Daltons while allowing removal of buffer salts and small contaminants. Membranes and cassettes are manufactured using clean-room conditions to ensure that are contaminant-free.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO):
Easy-loading—pipette-accessible for easy sample loading and retrieval
Self-floating—integrated air chambers eliminate the need for float buoys
Sturdy construction—ensures the highest possible sample integrity and protection
Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Versatile—ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting
Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination

Gamma-irradiated (sterile) dialysis cassettes
Gamma-irradiated Slide-A-Lyzer G2 Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post irradiation.

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 21%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionG2 Dialysis Cassettes
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO10 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)0.5 mL
Unit SizeEach
Contents & Storage
Store at room temperature

Frequently asked questions (FAQs)

If the Slide-A-Lyzer Dialysis Cassette is not reusable, why are there four portholes?

It is possible to add reagents to the Slide-A-Lyzer Dialysis Cassette at different times using different portholes, allow a reaction to occur, and then dialyze in the same Slide-A-Lyzer Dialysis Cassette, reducing sample loss through reduced sample handling.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How much sample will I lose in the Slide-A-Lyzer Dialysis Cassette?

The Slide-A-Lyzer Dialysis Cassette is the same regenerated cellulose as other dialysis tubing so you can expect about the same amount of protein loss as with tubing. As a guide, a 1 mg/mL solution will have a recovery rate of greater than 95%; at 100 mg/mL the recovery drops to 75-80%; and in solutions as dilute 10 µg/mL users may observe only a 50% (or less) recovery.

What conditions do you recommend for dialysis?

Dialyze for the amount of time sufficient to remove low molecular weight compounds for the specific downstream application. A typical dialysis procedure is as follows:

1) Dialyze for 2 hrs at room temperature or 4 degrees C.
2) Change the dialysis buffer and dialyze for another 2 hrs.
3) Change the dialysis buffer and dialyze overnight at 4 degrees C.

Use the dialysis buffer at 200-500 times the volume of the sample. For example, if the starting sample volume is 1 mL, then use a dialysis buffer amount that is equal to 200 mL-500 mL for each dialysis buffer change.

What is the buoy for?

The buoy plays two roles: it suspends the Slide-A-Lyzer Dialysis Cassette above the stirring bar during dialysis; it provides a stand to set the Slide-A-Lyzer Dialysis Cassette in on the bench-top

Is it necessary for me to buy needles and syringes if I already have them?

No, any 18 gauge needle at least one inch long, and syringes of sufficient volume will work.