Search Thermo Fisher Scientific
Catalog Number | Description | Volume (Metric) | Tip Style | Unit Size |
---|---|---|---|---|
5469 | ART SolventSafe 200, Filtered, Sterile, Rack | 200 μL | Micro Point | Case of 3840 |
5469PK also known as 5469 | ART SolventSafe 200, Filtered, Sterile, Rack | 200 μL | Micro Point | Pack of 384 |
5479E | ART SolventSafe 1000E, Filtered, Sterile, Rack | 1000 μL | Micro Point | Case of 3200 |
5479EPK also known as 5479E | ART SolventSafe 1000E, Filtered, Sterile, Rack | 1000 μL | Micro Point | Pack of 400 |
Perform applications consistently and accurately by using Thermo Scientific™ ART™ SolventSafe Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes