Nunc™ Biobanking and Cell Culture Cryogenic Tubes, 3.6mL, 72mm, internal thread - FAQs
View additional product information for Nunc™ Biobanking and Cell Culture Cryogenic Tubes, 3.6mL, 72mm, internal thread - FAQs (347643, 375353, 337516PK, 347627PK, 366524PK, 337516, 375418, 375418PK, 379189, 379146, 347597PK, 379146PK, 368632, 366656PK, 379189PK, 366656, 363401, 347597, 351934, 377224, 363401PK, 377267, 375353PK, 347643PK, 363452PK, 368632PK, 347627, 377224PK, 363452, 367997, 377267PK, 366524, 374080, 374081)
Error:This product does not have any FAQs available.