Sodium borohydride, 98%, Thermo Scientific Chemicals
Sodium borohydride, 98%, Thermo Scientific Chemicals
Sodium borohydride, 98%, Thermo Scientific Chemicals
Sodium borohydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium borohydride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 500 g
100 g
500 g
Catalog number 013432.57
also known as 013432-57
Price (USD)
801.00
Each
Add to cart
Quantity:
4 x 500 g
Request bulk or custom format
Price (USD)
801.00
Each
Add to cart
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (unspecified)≥97.5%
FormWhite
Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in the ammonia borane dehydrogenation. Sodium borohydride is used as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces the aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only