Search Thermo Fisher Scientific
![]() | Detecting low-abundance antigens with even the best conventional dye conjugates can be a challenge, as photobleaching can make it difficult to effectively observe and record staining. The exceptional photostability of Qdot probes conjugates can provide substantial benefits in the detection of low abundance targets. Due to their narrow and symmetric emission spectra, Qdot probes are also ideal for multicolor, multiplexed fluorescence detection using a single excitation source such as the 405 nm laser. |
Qdot streptavidin conjugates combine the unsurpassed photostability of Qdot probes with the highly specific binding properties of streptavidin. These powerful fluorescence detection reagents offer unique performance advantages in a wide variety of tissue labeling and flow cytometry experiments; they are efficiently excited using the 405 nm violet laser, and the Qdot probes fluorescence is extremely resistant to photobleaching.
These products provide 200 µL of the Qdot streptavidin conjugate and a detailed protocol outlining the use of the product in immunohistochemistry (IHC) applications using fluorescence microscopy. At conventional working concentrations, ~100 staining experiments may be performed
Each Qdot Streptavidin Sampler Kit provides 50 µL of each of six colors of Qdot streptavidin conjugate (525, 565, 585, 605, 655, and 705 nm emission) and a detailed protocol describing the use of the product in immunohistochemistry (IHC) applications using fluorescence microscopy. At conventional working concentrations, ~20 staining experiments with each color may be performed.
Qdot ITK probes are designed to allow researchers to create custom conjugations. Provided as carboxyl-derivatized, PEG-derivatized, and also as a version provided in decane (i.e., not water soluble)
Wheat germ agglutinin is a 36,000 molecular weight protein that binds to N-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues of glycoproteins and glycolipids. There are many uses for WGA conjugates including cell surface labeling and measuring retrograde neuronal transport. Our Qdot 655 wheat germ agglutinin conjugate provides highly sensitive labeling of N-acetylglucosamine residues, with low non-specific binding.
Qtracker cell labeling kits employ a custom targeting peptide to deliver intensely fluorescent Qdot probes into the cytoplasm of live cells in culture. The fluorescence can be traced through several generations and is not transferred to adjacent cells in a population.
Designed to deliver intense fluorescence, Qtracker vascular labels are non-targeted Qdot probes designed to be injected into the tail vein of mice for vasculature studies.