Search Thermo Fisher Scientific
Quickly measure the viscoelastic properties of your sample when you select the right rheometer or viscometer for your application.
Whether you measure texture consistency for new foods, analyze tensile strength of plastics, or flowability of paints and coatings, rely on our instruments to make life easier in the lab.
Quickly test viscosity and rheological properties of your materials for these industries:
Paints & Coatings • Processed Food • Cosmetics and Personal Care • Pharmaceuticals • Battery & Clean Energy • Polymers & Chemicals • Cement & Building Materials • Bitumen • Crude Oil
HAAKE Viscotester 3 | HAAKE Viscotester iQ/iQ Air | HAAKE MARS iQ/iQ Air | HAAKE MARS 40/60 | |
![]() | ![]() | ![]() | ![]() | |
Type of rheometer | handheld viscometer | rotational rheometer | rotational rheometer | rotational rheometer |
Portable | Yes | Yes | ||
Temperature control options | Yes | Yes | Yes | |
Automatic lift functionality | Yes | Yes | ||
Standalone operation | Yes | Yes | ||
Software control operation | Yes | Yes | Yes | |
Tests in stress controlled (CS) mode | Yes | Yes | Yes | |
Tests in oscillation mode (CD & CS) | iQ Air only | iQ Air only | Yes | |
Tests in controlled rate mode (CR) | Yes | Yes | Yes | Yes |
Normal force capabilities | Yes | Yes | ||
Min Rotational Speed (rpm) | 62.5 | 0.01a | 0.001b | 10-8c |
Maximum Rotational Speed (rpm) | 62.5 | 1500 | 2000 | 4500 |
Dimensions (L x W x H) | 180 x 90 x 175 mmd | 270 x 500 x 500 mm | 480 x 390 x 670 mm | 600 x 600 x 890 mm |
Software | No | RheoWin | RheoWin | RheoWin |
HAAKE Viscotester 3 | HAAKE Viscotester iQ/iQ Air | HAAAKE MARS iQ/iQ Air | HAAKE MARS 40/60 | |
![]() | ![]() | ![]() | ![]() | |
Pressure cells | Yes | Yes | Yes | |
Building material cell | Yes | Yes | Yes | |
Tribology | Yes | Yes | ||
UV curing | Yes | |||
Interfacial rheology | Yes | |||
Solid sample testing fixture for dynamical mechanical thermal analysis (DMTA) | Yes | |||
Sentmanat Extensional Rheometer Accessory (SER) | Yes | |||
Hyphenation Capabilities | Rheo-DEA* Rheo-Microscope Rheo-FTIR Rheo-Raman |
*Rheo-DEA: Rheological dialectrical analysis
HAAKE Viscotester 3 | HAAKE Viscotester iQ/iQ Air | HAAAKE MARS iQ/iQ Air | HAAKE MARS 40/60 | |
![]() | ![]() | ![]() | ![]() | |
Quality Control | Yes | Yes | Yes | Yes |
Research & Development | Limited | Limited | Yes | |
Single-point viscosity | Yes | Yes | Yes | Yes |
Absolute viscosity data | Yes | Yes | Yes | |
Flow/viscosity curve | Yes | Yes | Yes | |
Viscoelastic behavior | iQ Air only | iQ Air only | Yes | |
Polymer melt analysis | iQ Air only | Yes | ||
Low viscous samples (juices, dairy products, inks etc.) | Yes | iQ Air only | iQ Air only | Yes |
Composites | Yes |