Search Thermo Fisher Scientific
样本简介
本文采用QuEChERS样品前处理,结合液相色谱串联三重四极杆质谱平台建立了粮谷中12种真菌毒素的快速检测方法。前处理净化方法,操作简单,重复性好,回收率高。使用Accucure aQ表面多孔增强核色谱柱,增强了对极性化合物的保留及选择性,峰形良好。采用LC-MS/MS方法,15min内快速完成低浓度真菌毒素的分析,并获得较好的回收率,回收率范围60-110%,LOD为0.1-2 ug/kg,该方法完美适合于粮谷中多种真菌毒素的分析。
![]() | ![]() |