    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
Search button Close
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • TRA-1-81 Antibodies
          Invitrogen
          TRA-1-81 Monoclonal Antibody (TRA-1-81)
          4 References
          View all (9) TRA-1-81 antibodies
          Questions & Answers
          Cite TRA-1-81 Monoclonal Antibody (TRA-1-81)
          • Antibody Testing Data (7)
          TRA-1-81 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRA-1-81 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          TRA-1-81 Antibody (MA1-024) in ICC/IF
          Immunofluorescent analysis of TRA-1-81 using anti-TRA-1-81 monoclonal antibody (Product # MA1-024) shows staining on the cell surface of human H9 human embryonic stem cells, indicating pluripotency. TRA-1-81 staining (green) and an overlay image of TRA-1-81 with DAPI (blue) is shown. H9 human embryonic stem cells were grown on matrigel coated chamber slides and fixed with formaldehyde prior to staining. Cells were probed with a Monoclonal antibody recognizing TRA-1-81 (Product # MA1-024) at a dilution of 1:20 overnight at 4 °C, washed with... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRA-1-81 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-81 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-81 Antibody in Immunocytochemistry (ICC/IF)
          TRA-1-81 Antibody in Western Blot (WB)
          TRA-1-81 Antibody in Flow Cytometry (Flow)
          TRA-1-81 Antibody in Flow Cytometry (Flow)
          TRA-1-81 Antibody in Flow Cytometry (Flow)
          Product Details
          MA1-024
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:500
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          0.4 µg
          -
          Miscellaneous PubMed (MISC)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Human
          Published species
          Bovine, Human
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          TRA-1-81
          Immunogen
          human embryonal carcinoma cell line 2102Ep
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Affinity chromatography - MBP
          Storage buffer
          PBS with 1mg/mL BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536706
          Target Information
          TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0