Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For short term use, store at 2-8C up to 1 week. For long term storage, store at -20C in small aliquots to prevent freeze-thaw cycles.
Concentration is batch dependent: 0.5-1 mg/mL
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Controls development of mouthparts, and labial and maxillary palps.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG31481-PA; CG31481-PB; CG31481-PC; CG31481-PD; Homeotic protein proboscipedia; pb-PA; pb-PB; pb-PC; pb-PD; proboscipedia; proboscopedia; proposcipedia
Gene Aliases: BG:DS01719.1; CG17880; CG31481; Dmel\CG31481; Dmel_CG31481; Dmpb; l(3)04498; PB; pd; pr
UniProt ID: (Fruit fly) P31264
Entrez Gene ID: (Fruit fly) 40826
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support