Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVDKTPVMCS HFAQDVWPEH SIKDSFQKVI LRTYGKYGHE NLQLRKDHKS VDACKVYKGG YNGLNQCLTT TDSKIFQCDK YVKVFHKFPN VNRNKIRHTG KKPFKCKNRG KSFCMLSQLT QHKKIHTREY SYKCEECGKA FNWSSTLTKH KIIHTGEKPY KCEECGKAFN RSSNLTKHKI IHTGEKPYKC EECGKAFNRS STLTKHKRIH TEEKPYKCEE CGKAFNQFSI LNKHKRIHME DKPYKCEECG KAFRVFSILK KHKIIHTGEK PYKCEECGKA FNQFSNLTKH KIIHTGEKPY KCDECGKAFN QSSTLTKHKR IHTGEKPYKC EECGKAFKQS STLTEHKIIH TGEKPYKCEK CGKAFSWSSA FTKHKRNHME DKPYKCEECG KAFSVFSTLT KHKIIHTREK PYKCEECGKA FNRSSIFTKH KIIHTEGKSY KCEKCGNAFN QSSNLTARKI IYTGEKPYKY EECDKAFNKF STLITHQIIY TGEKPCKHEC GRAFNKSSNY TKEKLQT
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 203; Zinc finger protein 92; Zinc finger protein HTF12
Gene Aliases: HEL-203; HPF12; HTF12; TF12; ZNF92
UniProt ID: (Human) Q03936
Entrez Gene ID: (Human) 168374
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support