Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLNMQGAEER DIRRETCPGW VNKNKPALEQ DVCKIDSSGI VVKRFQEDEY QDSTFEEKYA CEGMKENSPR EIAESCLFQE GGFGRITFIH KEAPPEIISQ GYNFEKSLLL TSSLVTRLRV STEESLHQWE TSNIQTNDIS DQSKCPTLCT QKKSWKCNEC GKTFTQSSSL TQHQRTHTGE RPYTCEECGK AFSRSSFLVQ HQRIHTGVKP YGCEQCGKTF RCRSFLTQHQ RIHTGEKPYK CNECGNSFRN HSHLTEHQRI HTGEKPYKCN RCGKAFNQNT HLIHHQRIHT GEKPYICSEC GSSFRKHSNL TQHQRIHTGE KPHKCDECGK TFQTKANLSQ HQRIHSGEKP YKCKECGKAF CQSPSLIKHQ RIHTGEKPYK CKECGKAFTQ STPLTKHQRI HTGERPYKCS ECGKAFIQSI CLIRHQRSHT GEKPYKCNEC GKGFNQNTCL TQHMRIHTGE KPYKCKECGK AFAHSSSLTE HHRTHTGEKL YKCSECEKTF RKYAHLSEHY RIHTGEKPYE CIECGKFFRH SSVLFRHQKL HSGD
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 502
Gene Aliases: ZNF502
UniProt ID: (Human) Q8TBZ5
Entrez Gene ID: (Human) 91392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support